Learn More
Duran™ Keck Conical Ground Clamps
Made of temperature resistant polyacetal (POM)
Brand:  Duran™ 286336709
Product Code. 15971571
  • 3655.00 DKK / Pack of 100
Only null left
Estimated Shipment: 14-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • KECK™ Clips for conical joints are available in sizes NS 10 up to NS 45
  • Made of temperature resistant polyacetal (POM) Good spring properties
  • Colour coded by size
  • No risk of scratching the glass
  • Will not corrode or bend out of shape
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Duran™ Keck Conical Ground Clamps > Color: Orange; Size: 34.5 NS