Få mere at vide
KDOAM-25, MedChemExpress
Beskrivelse
KDOAM-25 is a potent and highly selective histone lysine demethylases 5 (KDM5) inhibitor with IC50s of 71 nM, 19 nM, 69 nM, 69 nM for KDM5A, KDM5B, KDM5C, KDM5D, respectively. KDOAM-25 increases global H3K4 methylation at transcriptional start sites and impairs proliferation in multiple myeloma MM1S cells.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | KDOAM-25 |
| CAS | 2230731-99-2 |
| Renhedsgrad noter | Research |
| Molekylær formel | C15H25N5O2 |
| Mængde | 5 mg |
| SMIL | NC(C1=CC=NC(CNCC(N(CC)CCN(C)C)=O)=C1)=O |
| Molekylvægt (g/mol) | 307.39 |
| Formel vægt | 307.39 |
| Grad | Research |
| Til brug med (applikation) | Cancer-programmed cell death |