missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ KDF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566984
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Immunogen sequence: ANWAKEHNGVP PSPDRAPPSR RDGQRLKSTM GSSFSYPDVK LKGIPVYPYP RATSPAPDAD SCCKEPLADP PPMRHSL Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
Plays a role in the regulation of the epidermis formation during early development. Required both as an inhibitor of basal cell proliferation and a promoter of differentiation of basal progenitor cell progeny.
Tekniske data
| KDF1 | |
| Polyclonal | |
| Unconjugated | |
| KDF1 | |
| 1810019J16Rik; C1orf172; C2H1orf172; KDF1; Keratinocyte differentiation factor 1; RGD1303271; RP11-344H11.3; uncharacterized protein C1orf172 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 126695 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NAX2 | |
| KDF1 | |
| Recombinant protein fragment of Human KDF1 (Product #RP-107629) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion