Learn More
Invitrogen™ KCNQ3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1930
Description
PA1-930 detects KCNQ3 from mouse, human and rat samples. This antibody is specific for KCNQ3 and does not detect KCNQ1, KCNQ2, KCNQ4 or KCNQ5. PA1-930 has been successfully used in immunofluorescence, immunohistochemistry and immunocytochemistry procedures. Immunohistochemical staining using PA1-930 yielded a strong signal mainly in interneurons and astrocytes in the dentate region of rat hippocampal samples. PA1-930 immunogen is a GST fusion protein encoding the first 71 amino acids of rat KCNQ3.
KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KCNQs (KCNQ1-5) found in the central nervous system and KCNQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.
Specifications
KCNQ3 | |
Polyclonal | |
Unconjugated | |
KCNQ3 | |
BFNC; BFNC2; EBN2; Kcnq3; KQT-like 3; KV7.3; potassium channel subunit alpha KvLQT3; potassium channel, voltage gated KQT-like subfamily Q, member 3; potassium channel, voltage-gated KQT-like subfamily Q, member 3; potassium channel, voltage-gated, subfamily Q, member 3; potassium voltage-gated channel subfamily KQT member 3; potassium voltage-gated channel subfamily Q member 3; potassium voltage-gated channel, KQT-like subfamily, member 3; potassium voltage-gated channel, subfamily Q, member 3; voltage-gated potassium channel subunit Kv7.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110862, 29682, 3786 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Frozen), Immunocytochemistry | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.02% sodium azide | |
O43525, O88944, Q8K3F6 | |
KCNQ3 | |
GST fusion protein encoding the first 71 amino acids of rat KCNQ3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.