Få mere at vide
KCNJ10 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-70371-0.025ML
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
KCNJ10 Polyclonal specifically detects KCNJ10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Tekniske data
| KCNJ10 | |
| Polyclonal | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| KCNJ10 | |
| A synthetic peptide from aa region 330-379 of human KCNJ10 conjugated to an immunogenic carrier protein was used as the antigen. The peptide is homologous in mouse and rat. | |
| 0.025 mL | |
| Primary | |
| Rat, mouse, human. Other species not yet tested. | |
| Mouse, Human, Rat | |
| Antisera |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| LYOPH | |
| Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| ATP-dependent inwardly rectifying potassium channel Kir4.1, ATP-sensitive inward rectifier potassium channel 10, BIRK-10, glial ATP-dependent inwardly rectifying potassium channel KIR4.1, Inward rectifier K(+) channel Kir1.2, inward rectifier K+ channel KIR1.2, KCNJ13-PEN, KIR1.2, Kir4.1, Potassium channel, inwardly rectifying subfamily J member 10, potassium inwardly-rectifying channel, subfamily J, member 10, SESAME | |
| Goat | |
| Unpurified | |
| RUO | |
| 3766 | |
| Reconstitute with 50 ul sterilized water to desired concetration. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion