missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KBTBD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05059-20UL
Additional Details : Weight : 0.00970kg
Description
KBTBD7 Polyclonal specifically detects KBTBD7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
KBTBD7 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
84078 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
DKFZP434E2318, kelch repeat and BTB (POZ) domain containing 7, kelch repeat and BTB domain-containing protein 7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 475-684 of human KBTBD7 (NP_115514.2). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |