Få mere at vide
Novus Biologicals™ KBTBD4 Overexpression Lysate
Artikelnummer. 18235702 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mg
Pakningsstørrelse:
0.1mg
Artikelnummer. Mængde Pakningsstørrelse
18235702 0.1 mg 0.1mg
1 options
Artikelnummer. 18235702 Leverandør Novus Biologicals™ Leverandørnr. NBL112132
 Begrænset vare
Forventet forsendelse: 15-10-2026
The KBTBD4 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Tekniske data
Cellelinje The lysate was created in HEK293T cells, using plasmid ID RC211572 and based on accession number NM_1895. The protein contains a C-terminal DDK tag.
Til brug med (applikation) Western Blot
Mængde 0.1 mg
Opbevaringsbuffer RIPA buffer
Renhed Protein
Testspecificitet Homo sapiens kelch repeat and BTB (POZ) domain containing 4 (KBTBD4), transcript variant 1, mRNA.
Indhold og opbevaring -80°C. Avoid freeze-thaw cycles.
Proteintilstand Native
For Research Use Only