missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
KATNAL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86689
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
KATNAL1 Polyclonal specifically detects KATNAL1 in Human samples. It is validated for Western Blot.
Tekniske data
| KATNAL1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84056 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| EC 3.6.4.3, katanin p60 ATPase-containing subunit A-like 1, katanin p60 subunit A-like 1MGC2599, p60 katanin-like 1 | |
| The immunogen is a synthetic peptide directed towards the middle region of human KATNAL1 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion