missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Reagent, for Karl Fischer Analysis, Modified, Solution ≈5mg H2O/mL
Click to view available options
Quantity:
500 mL
2.5 L
Specifications
Color | Brown |
Quantity | 500 mL |
Boiling Point | 124°C |
Packaging | Plastic Coated Glass bottle |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Karl Fischer reagent
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
- H370-Causes damage to organs.
- H372-Causes damage to organs.
- H360FD-May damage fertility. May damage the unborn child.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'