missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Reagent, for Karl Fischer Analysis, Modified, Solution ≈5mg H2O/mL
Click to view available options
Quantity:
500 mL
2.5 L
Specifications
Color | Brown |
Quantity | 2.5 L |
Boiling Point | 124°C |
Packaging | Plastic Coated Glass bottle |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'