missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Reagent, BAKER ANALYZED™, J.T. Baker™
Click to view available options
Quantity:
1 L
2.5 L
Description
- Solution Stabilized
- Contains Pyridine, 2-Methoxyethanol and Sulfur Dioxide
- One component reagent
Specifications
Flash Point | >21°C |
Quantity | 2.5 L |
UN Number | 1993 |
Concentration or Composition (by Analyte or Components) | 5.0 mg H2O/mL |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'