missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kanamycin A Deuterated (>90%), TRC
Specifications
Specifications
| Kemisk navn eller materiale | Kanamycin A Deuterated |
| CAS | 1174286-24-8 |
| Molekylær formel | C18H36N4O11 for undeuterated |
| Mængde | 1 mg |
| Synonym | O-3-Amino-3-deoxy-α-D-glucopyranosyl-(16)-O-[6-amino-6-deoxy-α-D-glucopyranosyl-(14)]-2-deoxy-D-streptamine Deuterated, Cantrex Deuterated, Cristalomicina Deuterated, Enterokanacin Deuterated, Kamycin Deuterated, Kanabristol Deuterated, Kanamytrex Deuterated, Kanaqua Deuterated, Kanasig Deuterated, Kanatrol Deuterated, Kanescin Deuterated, Otokalixin Deuterated, Resistomycin Deuterated |
| InChI formel | InChI=1S/C18H36N4O11/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17/h4-18,23-29H,1-3,19-22H2/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-/m1/s1 |
| SMIL | O[C@H]1[C@H](O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](CN)O2)[C@@H](N)C[C@@H](N)[C@@H]1O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O |
| Molekylvægt (g/mol) | 484.5 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?