Få mere at vide
MedChemExpress K114
Artikelnummer. 16450086
Encompass_Preferred
Skift visning
Mængde:
5 mg
100 mg
Pakningsstørrelse:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16450086 100 mg -
16446577 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16450086 Leverandør MedChemExpress Leverandørnr. HY103470
MedChemExpress K114, a fluorescent Congo Red analogue, binds tightly to amyloid fibrils with an EC50 of 20-30 nM. K114 is an efficient detector of semen-derived enhancer of virus infection (SEVI).
Tekniske data
Mængde 100 mg
Opbevaringskrav Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Opløselighed DMSO : 100 mg/mL (254.28 mM; Need ultrasonic)
Forsendelsestilstand Room temperature
CAS 872201-12-2
Molekylvægt (g/mol) 393.27 g/mol