Få mere at vide
Tocris Bioscience™ JZP 430
GREENER_CHOICE
Artikelnummer. 18769826
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18769826 10 mg
18739676 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18769826 Leverandør Tocris Bioscience™ Leverandørnr. 5844/10
Potent and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor
JZP 430 is a potent and irreversible α/β-hydrolase domain 6 (ABHD6) inhibitor (IC50 = 44 nM). Exhibits ∽230-fold selectivity over fatty acid amide hydrolase (FAAH) and lysosomal acid lipase (LAL).
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(4-Morpholinyl)-1,2,5-thiadiazol-3-yl N-cyclooctyl-N-methylcarbamate
CAS 1672691-74-5
Mængde 10 mg
Mål Hydrolase Inhibitors
Molekylær formel C16H26N4O3S
Formel vægt 354.47
Renhed >98%