Få mere at vide
Juvenile Hormone I, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Juvenile Hormone I |
| CAS | 13804-51-8 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C18H30O3 |
| Mængde | 0.25 mg |
| Synonym | [2R-[2α(2E,6E),3α]]-7-Ethyl-9-(3-ethyl-3-methyloxiranyl)-3-methyl-2,6-nonadienoic acid methyl ester, (2E,6E)-7-Ethyl-9-[(2R,3S)-3-ethyl-3-methyloxiranyl]-3-methyl-2,6-nonadienoic acid methyl ester, (E,E)-cis-10,11-Epoxy-7-ethyl-3,11-dimethyl-2,6-tridecadienoic acid methyl ester, (10R,11S)-(+)-Juvenile hormone I, (10R,11S)-Juvenile hormone I, (E,E)-cis-Juvenile hormone I, 10,11-Epoxy-7-ethyl-3,11-dimethyl-trans,trans-2,6-tridecadienoic acid methyl ester, 10,11-Epoxy-7-ethyl-3,11-dimethyltrideca-2-cis-6-dienoic acid methyl ester, AI 3-33972, AY 22243, C18-Juvenile hormone, Cecropia juvenile hormone, JH I, Juvenile hormone 1, Methyl (2E,6E,10Z)-10,11-epoxy-3,11-dimethyl-7-ethyl-2,6-tridecadienoate, Methyl (E,E cis)-10,11-epoxy-7-ethyl-3,11-dimethyl-2,6-tridecadienoate, Methyl (E,E)-3,11-dimethyl-7-ethyl-cis-10,11-epoxytrideca-2,6-dienoate, Methyl 10,11-epoxy-7-ethyl-3,11-dimethyl-2,6-tridecadienoate, Methyl 10,11-epoxy-7-ethyl-3,11-dimethyl-trans,trans,cis-2,6-tridecadienoate, Methyl 12,14-dihomojuvenate, Methyl 3,11-dimethyl-7-ethyl-cis-10,11-oxido-trans,trans-2,6-tridecadienoate, Methyl 7-ethyl-3,11-dimethyl-10,11-epoxy-2,6-tridecadienoate, Methyl cis-10,11-epoxy-3,11-dimethyl-7-ethyl-trans,trans-2,6-tridecadienoate, Methyl cis-10,11-epoxy-7-ethyl-3,11-dimethyl-trans,trans-2,6-tridecadienoate, Methyl trans,trans,cis-10,11-epoxy-7-ethyl-3,11-dimethyltrideca-2,6-dienoate, Methyl-12-homojuvenate, cis/trans-Methyl 10,11-epoxy-7-ethyl-3,11-dimethyl-2,6-tridecadienoate |
| InChI formel | InChI=1S/C18H30O3/c1-6-15(11-12-16-18(4,7-2)21-16)10-8-9-14(3)13-17(19)20-5/h10,13,16H,6-9,11-12H2,1-5H3/b14-13+,15-10+/t16-,18+/m1/s1 |
| SMIL | Cl.Clc1n(C)c2c(n1)c3c(nccc3)cc2N |
| Molekylvægt (g/mol) | 294.43 |
| Formel vægt | 294.2195 |
| Vis mere |