Få mere at vide
Juvenile Hormone B 3 (Mixture of Diastereomers), Technical Grade, TRC
Artikelnummer. 30136336
Skift visning
Mængde:
0.5 mg
2.5 mg
10 mg
Pakningsstørrelse:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136336 2.5 mg -
30134960 10 mg -
30009940 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136336 Leverandør Toronto Research Chemicals Leverandørnr. TRCJ2112052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120293-93-8
Molekylær formel C16H26O4
Molekylvægt (g/mol) 282.38
Synonym (2E)-5-[(2S,3S)-3-[2-[(2R)-3,3-Dimethyl-2-oxiranyl]ethyl]-3-methyl-2-oxiranyl]-3-methyl-2-pentenoic Acid Methyl Ester, (2E)-5-[(2S,3S)-3-[2-[(2R)-3,3-Dimethyloxiranyl]ethyl]-3-methyloxiranyl]-3-methyl-2-pentenoic Acid Methyl Ester, [2S-[2α(E),3β(S*)]]-5-[3-[2-(3,3-Dimethyloxiranyl)ethyl]-3-methyloxiranyl]-3-methyl-2-pentenoic Acid Methyl Ester, Methyl 6,7:10,11-Bis(epoxy)farnesoate
IUPAC navn methyl (E)-5-[(2S,3S)-3-[2-[(2R)-3,3-dimethyloxiran-2-yl]ethyl]-3-methyloxiran-2-yl]-3-methylpent-2-enoate
SMIL O=C(OC)/C=C(C)/CC[C@@H]1O[C@]1(CC[C@H]2OC2(C)C)C
Tekniske data
Kemisk navn eller materiale Juvenile Hormone B 3 (Mixture of Diastereomers)
Mængde 2.5 mg
InChI formel InChI=1S/C16H26O4/c1-11(10-14(17)18-5)6-7-13-16(4,20-13)9-8-12-15(2,3)19-12/h10,12-13H,6-9H2,1-5H3/b11-10+/t12-,13+,16+/m1/s1
Formel vægt 282.1831