Learn More
Justrite™ Antistatic Dual Alligator Clips
Dual alligator clips with end connectors
Brand:  Justrite™ 08500
Additional Details : Weight : 0.80000kg
Product Code. 11953549
  • 421.00 DKK / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
Compliance
Comply with OSHA standards and local safety codes requiring grounded containers during the transfer of liquids and drums connected to an earth ground.
Specifications
Specifications
Dual Alligator Clips
Zinc, Steel
36 in.
91.44 cm