Få mere at vide
3M™ Jupiter™ Calibration and Air Flow Indicator, 021-14-12P
Artikelnummer. 15959276 Shop alle 3M produkter
:
CE Approved
Artikelnummer. 15959276
Brand: 3M™ 7000104088
3M™ Jupiter™ Calibration and Air Flow Indicator enables the checking and calibration of 3M™ Jupiter™ Powered Air Turbo.
  • Ensure 3M™ Jupiter™ Powered Air Turbo is exceeding its minimum air flow rate
  • Ensure powered air turbo alarm point is correctly set
  • Air flow must be checked before using powered air turbo
  • Powered Air Turbo must be calibrated once a month
  • Accessory