missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JPS016
Description
JSP016 is a selective Degrader (PROTAC™) and inhibitor of class I HDACs (DC50 values are 530 nM and 550 nM for HDAC3 and HDAC1; Dmax is 77%, 66% and 45% at HDAC1, 3 and 2; IC50 values are 380 nM, 570 nM and 820 nM at HDAC3, 1 and 2). Comprises a benzamide-based compound joined by a linker to a Von Hippel Lindau (VHL) E3 ligase ligand. JPS016 treatment up- or down-regulates nearly 4000 differentially expressed genes, arrests cells at sub-G1 phase and promotes apoptosis in HCT116 cells.
Specifications
Specifications
| Kemisk navn eller materiale | (2S,4R)-1-((S)-2-(2-((9-(2-((4-((2-Aminophenyl)carbamoyl)phenyl)amino)-2-oxoethoxy)nonyl)oxy)acetamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 2669785-77-5 |
| Mængde | 5 mg |
| Mål | Active Degraders |
| Molekylær formel | C48H63N7O8S |
| Renhed | 0.97 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?