Få mere at vide
Tocris Bioscience™ JNJ 55511118
GREENER_CHOICE
Selective, high affinity negative modulator of AMPA receptors containing TARP-γ8
Brand:  Tocris Bioscience™ 6278/25
Encompass
Produktkode 15786379
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
JNJ 55511118 is a high affinity and selective negative modulator of AMPA receptors containing TARP-γ8 (Ki = 26 nM). Exhibits <50% binding against a panel of 52 receptors, ion channels and transporters (except 5HT2B and melatonin receptors 78% and 57%, respectively). Also displays minimal activity against other TARP-less AMPARs and AMPARs coexpressed with other TARPs or CNIH2. Orally bioavailable and brain penetrant. Anticonvulsant.
Tekniske data
Tekniske data
5-[2-Chloro-6-(trifluoromethoxy)phenyl]-1,3-dihydro-2H-benzimidazol-2-one
2036081-86-2
>98%
25 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner