Få mere at vide
Tocris Bioscience™ JNJ 55511118
Selective, high affinity negative modulator of AMPA receptors containing TARP-γ8
Brand: Tocris Bioscience™ 6278/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
JNJ 55511118 is a high affinity and selective negative modulator of AMPA receptors containing TARP-γ8 (Ki = 26 nM). Exhibits <50% binding against a panel of 52 receptors, ion channels and transporters (except 5HT2B and melatonin receptors 78% and 57%, respectively). Also displays minimal activity against other TARP-less AMPARs and AMPARs coexpressed with other TARPs or CNIH2. Orally bioavailable and brain penetrant. Anticonvulsant.Tekniske data
| 5-[2-Chloro-6-(trifluoromethoxy)phenyl]-1,3-dihydro-2H-benzimidazol-2-one | |
| 2036081-86-2 | |
| >98% | |
| 5 mg |