Få mere at vide
JNJ 42041935, TRC
Artikelnummer. 30133384
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133384 5 mg
30009465 10 mg
30007924 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133384 Leverandør Toronto Research Chemicals Leverandørnr. TRCJ2113185MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1193383-09-3
Molekylær formel C12 H6 Cl F3 N4 O3
Molekylvægt (g/mol) 346.65
Synonym 1-[6-Chloro-5-(trifluoromethoxy)-1H-benzimidazol-2-yl]-1H-pyrazole-4-carboxylic Acid, 1-(6-Chloro-5-(trifluoromethoxy)-1H-benzo[d]imidazol-2-yl)-1H-pyrazole-4-carboxylic Acid
IUPAC navn 1-[6-chloro-5-(trifluoromethoxy)-1H-benzimidazol-2-yl]pyrazole-4-carboxylic acid
SMIL OC(=O)c1cnn(c1)c2nc3cc(OC(F)(F)F)c(Cl)cc3[nH]2
Tekniske data
Kemisk navn eller materiale JNJ 42041935
Mængde 5 mg
InChI formel InChI=1S/C12H6ClF3N4O3/c13-6-1-7-8(2-9(6)23-12(14,15)16)19-11(18-7)20-4-5(3-17-20)10(21)22/h1-4H,(H,18,19)(H,21,22)
Formel vægt 346.0081