Få mere at vide
Tocris Bioscience™ JNJ 303
GREENER_CHOICE
Artikelnummer. 18014002
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18014002 10 mg
18043872 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18014002 Leverandør Tocris Bioscience™ Leverandørnr. 3899/10
Potent and selective IKs blocker
Potent IKs blocker (IC50 = 64 nM). Displays no effects on other cardiac channels; IC50 values are 3.3, >10, 11.1 and 12.6 μM for INa, ICa, Ito and IKr currents respectively. Prolongs QT and causes unprovoked torsades de pointes (TdP).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 878489-28-2
Molekylær formel C21H29ClN2O4S
Tekniske data
Kemisk navn eller materiale JNJ 303
Mængde 10 mg
Formel vægt 440.98
Renhed >98%
Molekylvægt (g/mol) 440.983