Få mere at vide
Tocris Bioscience™ JHW 007 hydrochloride
Artikelnummer. 18074462
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18074462 50 mg
18045432 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18074462 Leverandør Tocris Bioscience™ Leverandørnr. 4351/50
High affinity dopamine uptake inhibitor
Dopamine uptake inhibitor; displays high affinity for the dopamine transporter (DAT) (Ki = 25 nM compared to 1330 and 1730 for NET and SERT respectively). Suppresses the effects of cocaine administration in a dose-dependent manner and decreases cocaine and methamphetamine self-administration in rats.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202645-74-7
Molekylær formel C24H30ClF2NO
Tekniske data
Kemisk navn eller materiale JHW 007 hydrochloride
Mængde 50 mg
Formel vægt 421.95
Renhed >99%
Molekylvægt (g/mol) 421.957