Få mere at vide
Tocris Bioscience™ JH 295
GREENER_CHOICE
Irreversible Nek2 inhibitor
Brand:  Tocris Bioscience™ 4322/2
Encompass
Produktkode 15726339
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
JH 295 is an irreversible Nek2 inhibitor (IC50=770 nM). Inhibits Nek2 via alkylation of residue Cys22. Displays 5-fold selectivity for Nek2 over Cdk1. Exhibits no significant effect on Aurora B or Plk1 kinases, bipolar spindle assembly or spindle assembly checkpoint. Cell permeable.
Tekniske data
Tekniske data
N-[(3Z)-3-[(2-Ethyl-4-methyl-1H-imidazol-5-yl)methylene]-2,3-dihydro-2-oxo-1H-indol-5-yl]-2-propynamide
1311143-71-1
>98%
2 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner