Learn More
Chemsplash Jet Spray 88 Coverall Type 3B/4B 5B/6B with Feet

Jet Spray 88 Coverall with Feet Type 3B/4B 5B/6B is highly resistant against Chemical permeation offering high levels of protection against even the most corrosive of chemicals.
3895.00 DKK
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Gender | Unisex |
Material | Non-woven Fabric |
Color | Yellow |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
30058408
![]() |
Chemsplash
2617.700.004 |
Small |
3895.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058385
![]() |
Chemsplash
2617.700.005 |
Medium |
3895.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058346
![]() |
Chemsplash
2617.700.006 |
Large |
3895.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058174
![]() |
Chemsplash
2617.700.007 |
X-Large |
3895.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058206
![]() |
Chemsplash
2617.700.008 |
2X-Large |
3895.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
30058442
![]() |
Chemsplash
2617.700.009 |
3X-Large |
3895.00 DKK
Pack of 25 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
Description
Chemsplash Jet Spray 88 Coverall with Feet
- 88GSM Heavyweight Microporous Non Woven Fabric
- Ultrasonically Tape Welded Seams
- Self Adhesive Chin Strap for Optimum Protection
- Elasticated Hood, Half Waist, Cuffs and Ankles
- Convenient Thumb Loops at Wrist
- Single-Way Zip with Bi-Folding Self Adhesive Flap
- Silicone and Latex Free
- Non Linting Fabric
- Anti-Static
- PFAS Free
Description: Chemsplash Jet Spray 88 Coverall with Feet Type 3B/4B 5B/6B is highly resistant against chemical permeation, offering high levels of protection against even the most corrosive chemicals. Made from a thick 88gsm heavyweight polypropylene/polyethylene multilayer laminated material, it is also tested to provide protection against biological hazards to EN14126, offering protection against viruses, bacteria, and bloodborne pathogens.
Specifications
Cleanroom Coveralls | |
Non-woven Fabric | |
Single-use | |
Attached Boots | |
Hooded | |
Ties | |
Non-sterile | |
Yes | |
Class 100-100,000 (ISO 5-8) |
Unisex | |
Yellow | |
Elastic | |
Elastic, Thumb Loop | |
3-panel | |
Stitched, Taped | |
Liquid | |
None |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.