Få mere at vide
MAPA Professional Jersette 307 Chemical Protection Glove
Artikelnummer. 31749205
Skift visning
Størrelse:
6
7
8
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Størrelse
31749205 6
31749195 7
31749192 8
31749186 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 31749205 Leverandør MAPA Professional Leverandørnr. 34307016
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Chemical Protection Glove offering exceptional comfort and precision dexterity in light aggressive environments.
  • Main color: Mauve
  • Thickness (mm): 0.75
  • Length (mm): 310
  • ISO CUT Level: X
  • Risks: Heat - Liquid
  • Coating material: Grip-textured - Latex
  • Exceptional comfort, due to the precision of the textile support and the anatomical shape, Excellent dexterity and good grip.
  • Silicone free to prevent traces or defects prior to painting.
  • NORMS:
    • Category 2
    • EN 388: 2120X
    • EN 407: X1XXXX
Tekniske data
ANSI Abrasion Rating ANSI 0
Kloreret No
Applikationer Industrial
Farve Lavender
Fingertykkelse (metrisk) 0.75 mm
Latex fri No
Materiale Latex
Håndfladetykkelse (metrisk) 0.75 mm
Produkttype Latex Gloves
Størrelse 6
Tykkelse (metrisk) 0.75 mm
EN388 Standard EN388:2120X
EN 421 Yes
Handske manchet stil Scalloped
Greb design Smooth
Foring Seamless Lining