Learn More
Bibby Scientific™ Jenway™ Plastic Cuvettes For UV and Visible Region Wavelength
Product Code. 11475009
Change view
Click to view available options
Capacity (Metric):
0.07 to 1.5 mL
1.5 to 3.0 mL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11475009 0.07 to 1.5 mL Pack of 100
11430559 1.5 to 3.0 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11475009 Supplier Bibby Scientific™ Supplier No. 8307038
  • ONLINE EXCLUSIVE PRICE
    1875.00 DKK
    1514.39 DKK / Pakke med 100
    Spare 360.61 DKK
    19% Slukket
Forventet forsendelse: 21-12-2026
Save cleaning time and avoid cross-contamination with disposable plastic cuvettes. Bibby Scientific™ Jenway™ Plastic Cuvettes For UV and Visible Region Wavelength come in a pack of 100.
  • Efficient and time saving
  • Prevents cross-contamination
Specifications
Type Cuvette
Capacity (Metric) 0.07 to 1.5 mL
Material Plastic
Quantity 100/Pk.
Path Length 10 mm
For Use With (Application) Spectral Measurement of Liquids