Få mere at vide
Tocris Bioscience™ Jedi2
Beskrivelse
Jedi2 is a piezo1 channel activator. Promotes Ca2+ influx into HEK293 cells expressing Piezo1 channels. Elicits responses in mouse Piezo1-transfected, but not Piezo2-transfected cells.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Methyl-5-(2-thienyl)-3-furancarboxylic acid |
| CAS | 651005-90-2 |
| Mængde | 10 mg |
| Molekylær formel | C10H8O3S |
| Formel vægt | 208.23 |
| Renhed | 98% |