missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ Janelia Fluor™ 585, Haloalkane

Artikelnummer. 30245509
Skift visning
Klik for at se tilgængelige muligheder
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
30245509 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30245509 Leverandør Tocris Bioscience™ Leverandørnr. 8807/10U

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Cell-permeable fluorogenic fluorescent dye with a chloroalkane handle. Application: live-cell imaging as a self-labeling tag substrate

Key Information: Janelia Fluor™ 585, Haloalkane is an orange cell-permeable fluorogenic fluorescent dye with a chloroalkane handle.

Application: Live-cell imaging as a self-labeling tag substrate. Suitable for confocal microscopy, light sheet microscropy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells). Stable to fixation. Cell permeable.

Properties and Photophysical Data: Janelia Fluor™ 585, Haloalkane shows a high degree of fluorogenicity. Excitation and emission maxima (λ) are 585 nm and 609 nm, respectively; quantum yield = 0.78; extinction coefficient = 156,000 M-1cm-1 (measured in ethanol or TFE plus 0.1% TFA); lactone-zwitterion equilibrium constant (KL-Z) <0.0001. Turn-on ratio in vitro A/A0 = 79, F/F0 = 99. Exhibits low non-specific background staining.

Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 4-((2-(2-((6-Chlorohexyl)oxy)ethoxy)ethyl)carbamoyl)-2-(3-(3,3-difluoroazetidin-1-ium-1-ylidene)-6-(3,3-difluoroazetidin-1-yl)-10,10-dimethyl-3,10-dihydroanthracen-9-yl)benzoate
CAS 1811539-90-8
Mængde 10 μg
Mål Janelia Fluor Dyes
Molekylær formel C40H44ClF4N3O5
Renhed 95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.