missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ Janelia Fluor 549, SE

Beskrivelse
Janelia Fluor® 549, SE is a yellow fluorescent dye; supplied as an NHS ester for coupling to primary amine groups. NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Suitable for confocal fluorescent imaging and super resolution microscopy (SRM) techniques, such as dSTORM (live and fixed cells) and STED. Also suitable for flow cytometry. Cell permeable. Excitation maximum = 549 nm; emission maximum = 571 nm; Quantum yield = 0.88; Extinction coefficient = 101,000 M-1cm-1; A280 correction factor is 0.169.
We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals.
HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3,6-Di-1-azetidinyl-9-[2-carboxy-5-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]phenyl]xanthylium, inner salt |
| CAS | 1811539-32-8 |
| Synonym | JF 549, NHS |
| Mængde | 2 mg |
| Mål | Janelia Fluor Dyes |
| Molekylær formel | C31H25N3O7 |
| Formel vægt | 551.56 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?