missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAB1 Rabbit anti-Human, Mouse, Rat, Clone: 3X5F0, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16328-100UL
Additional Details : Weight : 0.00970kg
Description
JAB1 Monoclonal antibody specifically detects JAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
JAB1 | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
ChIP Assay, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3X5F0 | |
Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis), COP9 signalosome complex subunit 5, CSN5COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 5, EC 3.4, JAB138 kDa Mov34 homolog, Jun activation domain-binding protein 1, MOV-34, SGN5MGC3149, Signalosome subunit 5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human JAB1 (Q92905). | |
100 μg | |
Signal Transduction | |
10987 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |