Learn More
GPE Scientific™ J Young Spring Clips for Spherical Joints
GPE Scientific™ J Young Spring Clips for Spherical Joints are spring-based clips designed to securely fasten J Young spherical joints.
Brand:  GPE Scientific™ JC19
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 15804784
  • 206.00 DKK / Each
Estimated Shipment: 21-08-2024
to see stock.
Add to basket
Description
Description
  • Spring clip design
  • Securely fasten J Young spherical joints
Specifications
Specifications
Spring Clips for J Young Spherical Joints
18/7 and 18/9 spherical joints