Learn More
GPE Scientific™ J Young In Line Taps, Fine Thread
GPE Scientific™ J Young In Line Taps, Fine Thread are made in various stopcock sizes and with various side arm diameters.
Brand: GPE Scientific™ SPOR/8
This item is not returnable.
View return policy
Description
- In-line format tap
- Available in various stopcock sizes
- Fine thread type
- Available with different diameter side arms
Specifications
J Young in line tap, fine thread type | |
8 mm | |
ID: 8 mm, OD: 11 mm |
Product Content Correction
Product Title
GPE Scientific™ J Young In Line Taps, Fine Thread > Stopcock: 8mm
Spot an opportunity for improvement?Share a Content Correction