missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies IWP-4

Artikelnummer. 17190082
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17190082 10 mg
17180082 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17190082 Leverandør STEMCELL Technologies Leverandørnr. 72554

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

WNT pathway inhibitor; Inhibits Porcupine

  • Inhibitor of WNT Production-4 (IWP-4) inhibits WNT signaling by inactivating Porcupine, a membrane-bound O-acyltransferase responsible for palmitoylating WNT proteins, which is essential for WNT signaling ability and secretion (Chen et al.) IWP-4 impairs WNT pathway activity in vitro with an IC50 value of 25 nM (Chen et al.)
  • Promotes cardiomyocyte differentiation in human pluripotent stem cells after treatment with CHIR99021 (Lian et al.), 2012, 2013; Sequiera et al.)
  • Promotes cardiomyocyte differentiation in human embryonic stem cells following primitive streak induction with BMP4 and Activin A (Hudson et al.)

Tekniske data

CAS 686772-17-8
Synonym Inhibitor of Wnt Production-4
Mængde 10 mg
Molekylær formel C23H20N4O3S3
Renhed ≥95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.