Learn More
Ivermectin, TRC
Product Code. 30003530
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003530 1 g 1g
30040980 25 mg 25mg
18011250 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30003530 Supplier Toronto Research Chemicals Supplier No. TRCI9408001G
 Restricted Item
Forventet forsendelse: 03-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70288-86-7
Molecular Formula C48 H74 O14 . C47 H72 O14
Molecular Weight (g/mol) 1736.16
Synonym 22,23-Dihydroavermectin B1, Bagomectina, Bimectin, Bovinet, Cardomec, Cardotek 30, Cardotek Plus, Cevamec, Equimec, Equimec Paste, Equimectrin, Equirat Gold, Eqvalan, Heartgard, Heartgard 30, Hyvermectin, Ivectin, Ivermec, Iversalt, Ivertin, Ivomec, Ivomec Gold, Ivosint, Jetamec, L 640471, MK 933, Mectizan, Mukhnet I, Noramectin, Noromectin, Oramec, Oramectin, Pandex, Phoenectin, Solantra, Stromectol, Uvemec, Vermic, Vetmec, Virbamec LA, Zimecterin
IUPAC Name Ivermectin
SMILES CC[C@H](C)[C@H]1O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2.CO[C@H]8C[C@H](O[C@H]9[C@H](C)O[C@H](C[C@@H]9OC)O[C@H]%10[C@@H](C)\C=C\C=C\%11/CO[C@@H]%12[C@H](O)C(=C[C@@H](C(=O)O[C@H]%13C[C@@H](C\C=C\%10/C)O[C@@]%14(CC[C@H](C)[C@H](O%14)C(C)C)C%13)[C@]%11%12O)C)O[C@@H](C)[C@@H]8O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C48H74O14.C47H72O14/c1-11-25(2)43-28(5)17-18-47(62-43)23-34-20-33(61-47)16-15-27(4)42(26(3)13-12-14-32-24-55-45-40(49)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-10)44(31(8)57-39)60-38-21-36(53-9)41(50)30(7)56-38;1-24(2)41-27(5)16-17-46(61-41)22-33-19-32(60-46)15-14-26(4)42(25(3)12-11-13-31-23-54-44-39(48)28(6)18-34(45(50)57-33)47(31,44)51)58-38-21-36(53-10)43(30(8)56-38)59-37-20-35(52-9)40(49)29(7)55-37/h12-15,19,25-26,28,30-31,33-45,49-50,52H,11,16-18,20-24H2,1-10H3;11-14,18,24-25,27,29-30,32-44,48-49,51H,15-17,19-23H2,1-10H3/b13-12+,27-15+,32-14+;12-11+,26-14+,31-13+/t25-,26-,28-,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41-,42-,43+,44-,45+,47+,48+;25-,27-,29-,30-,32+,33-,34-,35-,36-,37-,38-,39+,40-,41+,42-,43-,44+,46+,47+/m00/s1
Formula Weight 1735.0001
Chemical Name or Material Ivermectin