Få mere at vide
Ivermectin B1 Monosaccharide, TRC
Artikelnummer. 30138012
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138012 500 mg
30010514 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138012 Leverandør Toronto Research Chemicals Leverandørnr. TRCI940810500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 71837-27-9
Molekylær formel C41 H62 O11
Molekylvægt (g/mol) 730.92
Synonym Avermectin A1a, 4'-O-de(2,6-dideoxy-3-O-methyl-α-L-arabino-hexopyranosyl)-5-O-demethyl-22,23-dihydro-
SMIL CC[C@H](C)[C@H]1O[C@]2(CC[C@@H]1C)C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O)[C@H](C)O4)[C@@H](C)\C=C\C=C\5/CO[C@@H]6[C@H](O)C(=C[C@@H](C(=O)O3)[C@]56O)C)O2
Tekniske data
Kemisk navn eller materiale Ivermectin B1 Monosaccharide
Mængde 500 mg
InChI formel InChI=1S/C41H62O11/c1-9-22(2)37-25(5)15-16-40(52-37)20-30-18-29(51-40)14-13-24(4)36(50-33-19-32(46-8)35(43)27(7)48-33)23(3)11-10-12-28-21-47-38-34(42)26(6)17-31(39(44)49-30)41(28,38)45/h10-13,17,22-23,25,27,29-38,42-43,45H,9,14-16,18-21H2,1-8H3/b11-10+,24-13+,28-12+/t22-,23-,25-,27-,29+,30-,31-,32-,33-,34+,35-,36-,37+,38+,40+,41+/m0/s1
Formel vægt 730.4292