Få mere at vide
Itraconazole-d3, TRC
Artikelnummer. 30406351
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30406351 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406351 Leverandør Toronto Research Chemicals Leverandørnr. TRCI9375035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Itraconazole-d3
CAS 1217512-35-0
Molekylær formel C35 2H3 H35 Cl2 N8 O4
Mængde 5 mg
Synonym Itraconazole-d3
InChI formel InChI=1S/C35H38Cl2N8O4/c1-3-25(2)45-34(46)44(24-40-45)29-7-5-27(6-8-29)41-14-16-42(17-15-41)28-9-11-30(12-10-28)47-19-31-20-48-35(49-31,21-43-23-38-22-39-43)32-13-4-26(36)18-33(32)37/h4-13,18,22-25,31H,3,14-17,19-21H2,1-2H3/t25?,31-,35-/m0/s1/i2D3
SMIL [2H]C([2H])([2H])C(CC)N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(OC[C@H]5CO[C@@](Cn6cncn6)(O5)c7ccc(Cl)cc7Cl)cc4
IUPAC navn 4-[4-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-(1,1,1-trideuteriobutan-2-yl)-1,2,4-triazol-3-one
Molekylvægt (g/mol) 708.65
Formel vægt 707.2581