missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITIH5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-09495-100UL
Additional Details : Weight : 0.00970kg
Description
ITIH5 Polyclonal specifically detects ITIH5 in Human samples. It is validated for Western Blot.Specifications
ITIH5 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
inter-alpha (globulin) inhibitor H5, inter-alpha-inhibitor heavy chain 5, inter-alpha-trypsin inhibitor heavy chain H5, ITI heavy chain H5, ITIH5 inter-alpha-trypsin inhibitor heavy chain family, member 5, ITI-HC5, PP14776 | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human ITIH5 (NP_085046). Peptide sequence AAFITNFTMLIGDKVYQGEITEREKKSGDRVKEKRNKTTEENGEKGTEIF | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
80760 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |