missing translation for 'onlineSavingsMsg'
Learn More

Isoxanthohumol, TRC

Product Code. 30406101
Change view
Click to view available options
Mængde:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30406101 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406101 Supplier Toronto Research Chemicals Supplier No. TRCI9182132.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Isoxanthohumol
CAS 521-48-2
Molekylær formel C21H22O5
Mængde 2.5 mg
Synonym 2,3-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-5-methoxy-8-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one, 4',7-Dihydroxy-5-methoxy-8-(3-methyl-2-butenyl)-flavanone
InChI formel InChI=1S/C21H22O5/c1-12(2)4-9-15-16(23)10-19(25-3)20-17(24)11-18(26-21(15)20)13-5-7-14(22)8-6-13/h4-8,10,18,22-23H,9,11H2,1-3H3
SMIL COc1cc(O)c(CC=C(C)C)c2OC(CC(=O)c12)c3ccc(O)cc3
Molekylvægt (g/mol) 354.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.