Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Isoxaflutole |
| Emballage | Glass Bottle |
| CAS | 141112-29-0 |
| Farve | White |
| Smeltepunkt | 140°C |
| Kogepunkt | Decomposes below the boiling point. |
| Synonym | (5-cyclopropyl-4-isoxazolyl)[2-(methylsulfonyl)-4-(trifluoromethyl)phenyl]methanone |
| Industry Type | Environmental Analysis |
| Molekylvægt (g/mol) | 359.32 |
| Mængde | 100 mg |
| Vis mere |