missing translation for 'onlineSavingsMsg'
Learn More

Isoxaflutole 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30324521
Change view
Click to view available options
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30324521 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324521 Supplier Dr. Ehrenstorfer Supplier No. DREA14481000AC1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Isoxaflutole
CAS 141112-29-0
Analyt- eller komponentnavne Isoxaflutole
Forsendelsestilstand Room Temperature
Molekylær formel C15 H12 F3 N O4 S
Koncentration 1000 μg/mL
Certificeringer/compliance ISO 17034
Synonym (5-Cyclopropyl-4-isoxazolyl)[2-(methylsulfonyl)-4-(trifluoromethyl)phenyl]methanone, Balance, Balance (herbicide), Balance Pro, EXP 30953, EXP 31130A, Isoxaflutole, Merlin, Provence, RP 201772, RPA 201772, Methanone, (5-cyclopropyl-4-isoxazolyl)[2-(methylsulfonyl)-4-(trifluoromethyl)phenyl]-
InChI formel InChI=1S/C15H12F3NO4S/c1-24(21,22)12-6-9(15(16,17)18)4-5-10(12)13(20)11-7-19-23-14(11)8-2-3-8/h4-8H,2-3H2,1H3
SMIL CS(=O)(=O)c1cc(ccc1C(=O)c2cnoc2C3CC3)C(F)(F)F
IUPAC navn (5-cyclopropyl-1,2-oxazol-4-yl)-[2-methylsulfonyl-4-(trifluoromethyl)phenyl]methanone
Opløsningsmiddel eller Matrix Acetone
Molekylvægt (g/mol) 359.32
Mængde 1 ml
Formel vægt 359.0439 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.