Få mere at vide
Isovaleric Acid-d9, TRC
Artikelnummer. 30404823
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30404823 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404823 Leverandør Toronto Research Chemicals Leverandørnr. TRCI917572250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isovaleric Acid-d9
CAS 344298-81-3
Molekylær formel C5 2H9 H O2
Mængde 250 mg
Synonym Butanoic-2,2,3,4,4,4-d6 acid, 3-(methyl-d3)- (9CI), 3-Methylbutyric-d9 acid, Isovaleric acid-d9, 3-(Methyl-d3)butanoic-2,2,3,4,4,4-d6 acid, 3-Methylbutanoic-D9 acid, 3-Methylbutanoic acid-d9
InChI formel InChI=1S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)/i1D3,2D3,3D2,4D
SMIL [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,4,4,4-hexadeuterio-3-(trideuteriomethyl)butanoic acid
Molekylvægt (g/mol) 111.19
Formel vægt 111.1246