missing translation for 'onlineSavingsMsg'
Få mere at vide

(-)-Isosamidin, TRC

Artikelnummer. 30408837
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30408837 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408837 Supplier Toronto Research Chemicals Supplier No. TRCS1105051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (-)-Isosamidin
CAS 53023-18-0
Molekylær formel C21 H22 O7
Mængde 1 mg
Synonym 3-Methyl-2-butenoic acid (9R,10R)-9-(acetyloxy)-9,10-dihydro-8,8-dimethyl-2-oxo-2H,8H-benzo[1,2-b:3,4-b']dipyran-10-yl Ester, (9R-cis)-3-methyl-2-Butenoic Acid 9-(Acetyloxy)-9,10-dihydro-8,8-dimethyl-2-oxo-2H,8H-benzo[1,2-b:3,4-b']dipyran-10-yl Ester, Isosamidin, 3'(R),4'(R)-3'-Acetoxy-4'-senecioyloxy-3',4'-dihydroseselin
InChI formel InChI=1S/C21H22O7/c1-11(2)10-16(24)27-19-17-14(28-21(4,5)20(19)25-12(3)22)8-6-13-7-9-15(23)26-18(13)17/h6-10,19-20H,1-5H3/t19-,20-/m1/s1
SMIL CC(=CC(=O)O[C@H]1[C@@H](OC(=O)C)C(C)(C)Oc2ccc3C=CC(=O)Oc3c12)C
IUPAC navn [(9R,10R)-9-acetoxy-8,8-dimethyl-2-oxo-9,10-dihydropyrano[2,3-f]chromen-10-yl] 3-methylbut-2-enoate
Molekylvægt (g/mol) 386.4
Formel vægt 386.1366
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.