Få mere at vide
Isorhynchophylline, TRC
Artikelnummer. 30133940
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133940 25 mg
30012542 10 mg
30008646 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133940 Leverandør Toronto Research Chemicals Leverandørnr. TRCI90071025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6859-01-4
Molekylær formel C22 H28 N2 O4
Molekylvægt (g/mol) 384.47
Synonym Spiro[3H-indole-3,1'(5'H)-indolizine]-7'-acetic acid, 6'-ethyl-1,2,2',3',6',7',8',8'a-octahydro-α-(methoxymethylene)-2-oxo-, methyl ester, (αE,1'S,6'R,7'S,8'aS)-, 7-Isorhyncophylline (8CI), Corynoxan-16-carboxylic acid, 16,17-didehydro-17-methoxy-2-oxo-, methyl ester, (16E,20α)-, Isorhyncophylline (6CI,7CI), Spiro[3H-indole-3,1'(5'H)-indolizine]-7'-acetic acid, 6'-ethyl-1,2,2',3',6',7',8',8'a-octahydro-α-(methoxymethylene)-2-oxo-, methyl ester, [1'S-[1'α,6'α,7'β(E),8'aα]]-, Isorhynchophylline
IUPAC navn methyl (E)-2-[(3S,6'R,7'S,8'aS)-6'-ethyl-2-oxospiro[1H-indole-3,1'-3,5,6,7,8,8a-hexahydro-2H-indolizine]-7'-yl]-3-methoxyprop-2-enoate
SMIL CC[C@H]1CN2CC[C@]3([C@@H]2C[C@@H]1\C(=C/OC)\C(=O)OC)C(=O)Nc4ccccc34
Tekniske data
Kemisk navn eller materiale Isorhynchophylline
Mængde 25 mg
InChI formel InChI=1S/C22H28N2O4/c1-4-14-12-24-10-9-22(17-7-5-6-8-18(17)23-21(22)26)19(24)11-15(14)16(13-27-2)20(25)28-3/h5-8,13-15,19H,4,9-12H2,1-3H3,(H,23,26)/b16-13+/t14-,15-,19-,22-/m0/s1
Formel vægt 384.2049