missing translation for 'onlineSavingsMsg'
Learn More
Isorhamnetin-3-O-glucoside, MedChemExpress Product Code.: 16431508

Isorhamnetin-3-O-glucoside, MedChemExpress

Product Code. p-200100224
5 mg, 5mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16431508

Brand: Medchem Express HYN0777

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isorhamnetin-3-O-glucoside, a natural compound widely contained in many vegetables and rice, could be metabolized in intestinal microbiota after digestion.

Isorhamnetin-3-O-glucoside, a natural compound widely contained in many vegetables and rice, could be metabolized in intestinal microbiota after digestion.

Specifications

Chemical Name or Material Isorhamnetin-3-O-glucoside
CAS 5041-82-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H22O12
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (522.58 mM; Need ultrasonic)
SMILES O=C1C(O[C@@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO)=C(C3=CC(OC)=C(O)C=C3)OC4=CC(O)=CC(O)=C14
Molecular Weight (g/mol) 478.4
Formula Weight 478.4
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Isorhamnetin-3-O-glucoside, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.