Få mere at vide
Isoquinoline N-oxide, 98%, Thermo Scientific™
Artikelnummer. 11428627
Skift visning
Mængde:
5 g
25 g
100 g
Pakningsstørrelse:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11428627 100 g 100g
11408627 5 g 5g
11418627 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11428627 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A19738.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1532-72-5
Molekylær formel C9H7NO
Molekylvægt (g/mol) 145.161
MDL nummer MFCD00006903
InChI nøgle RZIAABRFQASVSW-UHFFFAOYSA-N
Synonym isoquinoline n-oxide, isoquinoline 2-oxide, isoquinoline-n-oxide, isoquinoline oxide, isoquinoline, 2-oxide, isoquinolin-n-oxide, isoquinolin-2-ium-2-olate, isoquinolinen-oxide, acmc-1boxv, 2-oxido-isoquinolin-2-ium
PubChem CID 290378
IUPAC navn 2-oxidoisoquinolin-2-ium
SMIL C1=CC=C2C=[N+](C=CC2=C1)[O-]
Tekniske data
Kemisk navn eller materiale Isoquinoline N-oxide
Smeltepunkt 103°C to 105°C
Kogepunkt 170°C (3 mmHg)
Mængde 100 g
Beilstein 114345
Følsomhed Hygroscopic
Formel vægt 145.16
Procent renhed 98%