Få mere at vide
Isoquinoline-d7, CDN
Artikelnummer. 30320719
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320719 0.25 g
30321842 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320719 Leverandør CDN Leverandørnr. CDND09040.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isoquinoline-d7
CAS 17157-12-9
Molekylær formel C9 D7 N
Mængde 0.25 g
Synonym Isoquinoline-1,3,4,5,6,7,8-d7 (ACI), Isoquinoline-d7 (8 CI, 9 CI)
InChI formel InChI=1 S/C9H7N/c1-2-4-9-7-10-6-5-8(9)3-1/h1-7 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D
SMIL [2 H]c1nc([2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c2c1[2 H]
IUPAC navn 1,3,4,5,6,7,8-heptadeuterioisoquinoline
Molekylvægt (g/mol) 136.2
Formel vægt 136.1018 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity