missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl trifluoroacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-7023599
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
400-38-4
C5H7F3O2
156.10
MFCD00013566
ASAXRKSDVDALDT-UHFFFAOYSA-N
isopropyl trifluoroacetate, isopropyltrifluoroacetate, isopropyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, 1-methylethyl ester, iso-propyl trifluoroacetate, trifluoroacetic acid isopropyl ester, pubchem12590, acmc-1ctsh, methylethyl 2,2,2-trifluoroacetate
78998
propan-2-yl 2,2,2-trifluoroacetate
CC(C)OC(=O)C(F)(F)F
This item is not returnable. View return policy
400-38-4
C5H7F3O2
156.10
MFCD00013566
ASAXRKSDVDALDT-UHFFFAOYSA-N
isopropyl trifluoroacetate, isopropyltrifluoroacetate, isopropyl 2,2,2-trifluoroacetate, acetic acid, trifluoro-, 1-methylethyl ester, iso-propyl trifluoroacetate, trifluoroacetic acid isopropyl ester, pubchem12590, acmc-1ctsh, methylethyl 2,2,2-trifluoroacetate
78998
propan-2-yl 2,2,2-trifluoroacetate
CC(C)OC(=O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point <-78°C
Density 1.108
Boiling Point 73°C
Flash Point −8°C (17°F)
Refractive Index 1.319
Quantity 25 g
UN Number UN2924
Beilstein 1764312
Sensitivity Moisture sensitive
Formula Weight 156.1
Percent Purity 97%
Chemical Name or Material Isopropyl trifluoroacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'