Learn More
Isopropyl Acetate, Spex CertiPrep
Product Code. 11934904 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1.2 mL
Unit Size:
1mL
Product Code. Quantity Unit Size
11934904 1.2 mL 1mL
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 11934904 Supplier SPEX Certiprep Supplier No. S2305
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Isopropyl Acetate Organic Single in Methanol-P&T, 1 mL, 1000 μg/mL, SPEX CertiPrep™
  • Isopopyl Acetate
  • CAS #108-21-4
  • 1 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Specifications
Packaging Ampule
CAS 108-21-4
Physical Form Liquid
Molecular Formula C5H10O2
Technique AA, ICP
InChI Key JMMWKPVZQRWMSS-UHFFFAOYSA-N
Type Certified Reference Material
SMILES CC(C)OC(=O)C
IUPAC Name propan-2-yl acetate
Molecular Weight (g/mol) 102.133
PubChem CID 7915
Quantity 1.2 mL
Chemical Name or Material Isopropyl Acetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Isopropyl acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol